Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001128591.1 | 5240 | Intron | NP_001122063.1 | ||
NM_001128592.1 | 5240 | Intron | NP_001122064.1 | ||
NM_001135750.1 | 5240 | Intron | NP_001129222.1 | ||
XM_011514591.2 | 5240 | Intron | XP_011512893.1 | ||
XM_011514593.2 | 5240 | Intron | XP_011512895.1 | ||
XM_011514594.2 | 5240 | Intron | XP_011512896.1 | ||
XM_011514595.2 | 5240 | Intron | XP_011512897.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001286455.1 | 5240 | UTR 3 | NP_001273384.1 | ||
NM_001286456.1 | 5240 | Intron | NP_001273385.1 | ||
NM_015482.1 | 5240 | UTR 3 | NP_056297.1 | ||
NM_021945.6 | 5240 | UTR 3 | NP_068764.3 | ||
XM_011514801.2 | 5240 | UTR 3 | XP_011513103.1 | ||
XM_017011180.1 | 5240 | UTR 3 | XP_016866669.1 | ||
XM_017011181.1 | 5240 | UTR 3 | XP_016866670.1 | ||
XM_017011182.1 | 5240 | UTR 3 | XP_016866671.1 | ||
XM_017011183.1 | 5240 | Intron | XP_016866672.1 | ||
XM_017011184.1 | 5240 | UTR 3 | XP_016866673.1 | ||
XM_017011185.1 | 5240 | Intron | XP_016866674.1 | ||
XM_017011186.1 | 5240 | UTR 3 | XP_016866675.1 |