Product Details

SNP ID
rs1207582
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:24363818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCAGGCCTGGGACACCAAGCCAA[A/G]CATCATGCAGTCGTTTAATGATCCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SUPT20HL1 PubMed Links

Gene Details

Gene
SUPT20HL1
Gene Name
SPT20 homolog, SAGA complex component-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136234.1 1058 Missense Mutation AAC,AGC N353S NP_001129706.1

View Full Product Details