Product Details

SNP ID
rs1211442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:84670481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCCCGGCGCGCCGGGCCATGCA[G/T]CGACGGCCGCCGCGGAGCTCCGAGC
Phenotype
MIM: 600456
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NTRK2 PubMed Links

Gene Details

Gene
NTRK2
Gene Name
neurotrophic receptor tyrosine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007097.2 645 UTR 5 NP_001007098.1
NM_001018064.2 645 UTR 5 NP_001018074.1
NM_001018065.2 645 UTR 5 NP_001018075.1
NM_001018066.2 645 UTR 5 NP_001018076.1
NM_001291937.1 645 UTR 5 NP_001278866.1
NM_006180.4 645 UTR 5 NP_006171.2
XM_005252001.2 645 UTR 5 XP_005252058.1
XM_005252003.2 645 UTR 5 XP_005252060.1
XM_005252004.2 645 UTR 5 XP_005252061.1
XM_005252006.3 645 UTR 5 XP_005252063.1
XM_005252007.3 645 UTR 5 XP_005252064.1
XM_011518718.2 645 UTR 5 XP_011517020.1
XM_011518720.2 645 UTR 5 XP_011517022.1
XM_017014751.1 645 UTR 5 XP_016870240.1
XM_017014752.1 645 UTR 5 XP_016870241.1
XM_017014753.1 645 UTR 5 XP_016870242.1
XM_017014754.1 645 UTR 5 XP_016870243.1
XM_017014755.1 645 UTR 5 XP_016870244.1
XM_017014756.1 645 UTR 5 XP_016870245.1
XM_017014757.1 645 UTR 5 XP_016870246.1
XM_017014758.1 645 UTR 5 XP_016870247.1
XM_017014759.1 645 UTR 5 XP_016870248.1
XM_017014760.1 645 UTR 5 XP_016870249.1
XM_017014761.1 645 UTR 5 XP_016870250.1

View Full Product Details