Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_014384.2 | Intron | NP_055199.1 | |||
XM_005271505.3 | Intron | XP_005271562.1 | |||
XM_011542750.2 | Intron | XP_011541052.1 | |||
XM_017017542.1 | Intron | XP_016873031.1 | |||
XM_017017543.1 | Intron | XP_016873032.1 | |||
XM_017017544.1 | Intron | XP_016873033.1 | |||
XM_017017545.1 | Intron | XP_016873034.1 | |||
XM_017017546.1 | Intron | XP_016873035.1 | |||
XM_017017547.1 | Intron | XP_016873036.1 | |||
XM_017017548.1 | Intron | XP_016873037.1 | |||
XM_017017549.1 | Intron | XP_016873038.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001037304.1 | Intron | NP_001032381.1 | |||
NM_001037305.1 | Intron | NP_001032382.1 | |||
NM_014174.2 | Intron | NP_054893.1 | |||
NM_199297.1 | Intron | NP_954994.1 | |||
NM_199298.1 | Intron | NP_954995.1 | |||
XM_005271527.3 | Intron | XP_005271584.1 | |||
XM_005271528.3 | Intron | XP_005271585.1 |