Product Details

SNP ID
rs1044383
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:78615194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATCCAAGTTCTGAAGACAAGTG[A/G]GGCTTATCAACAGGCCACTGTATCT
Phenotype
MIM: 615906
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNF219 PubMed Links

Gene Details

Gene
RNF219
Gene Name
ring finger protein 219
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024546.3 2626 UTR 3 NP_078822.3
XM_011535225.1 2626 UTR 3 XP_011533527.1
XM_017020743.1 2626 UTR 3 XP_016876232.1
Gene
RNF219-AS1
Gene Name
RNF219 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details