Product Details

SNP ID
rs1364704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:119217545 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAAGAAATTTCAAATTGAGGTGC[A/C]TTTGGCTATTTTATACATAATTATT
Phenotype
MIM: 609684
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MAL2 PubMed Links

Gene Details

Gene
MAL2
Gene Name
mal, T-cell differentiation protein 2 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052886.2 Intron NP_443118.1
XM_011516807.2 Intron XP_011515109.1

View Full Product Details