Product Details

SNP ID
rs1670177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123251961 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAGTGCCTGGTACATATTAAGCTC[C/T]CTATAAATAATTGTTAAAATAAATA
Phenotype
MIM: 604764
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZHX1 PubMed Links

Gene Details

Gene
ZHX1
Gene Name
zinc fingers and homeoboxes 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017926.2 Intron NP_001017926.1
NM_007222.4 Intron NP_009153.3
Gene
ZHX1-C8orf76
Gene Name
ZHX1-C8orf76 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204180.1 Intron NP_001191109.1

View Full Product Details