Product Details

SNP ID
rs1049402
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:30595045 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCGGTCCCTCAGCGCGGCCTCCTGC[C/G]CCCCGATCTCCTTGCCCGCCGCCGC
Phenotype
MIM: 600287
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GARS PubMed Links

Gene Details

Gene
GARS
Gene Name
glycyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316772.1 311 UTR 5 NP_001303701.1
NM_002047.3 311 Missense Mutation CCC,GCC P42A NP_002038.2
XM_006715686.2 311 UTR 5 XP_006715749.1

View Full Product Details