Product Details
- SNP ID
-
rs1483570
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:78674220 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TACGTGGTTTTTCTATGTGCTGAAT[C/T]CATTTTTGTCAATAGTTAATTTTTA
- Phenotype
-
MIM: 146660
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
IL7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78895468] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- IL7
- Gene Name
- interleukin 7
There are no transcripts associated with this gene.
- Gene
- ZC2HC1A
- Gene Name
- zinc finger C2HC-type containing 1A
View Full Product Details