Product Details

SNP ID
rs4763
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:45394123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGAGCACAGCACCTGGCACAGA[A/G]GAGCAGACAATAAATACCGGCTGTG
Phenotype
MIM: 610591
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP27 PubMed Links

Gene Details

Gene
ARHGAP27
Gene Name
Rho GTPase activating protein 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282290.1 4403 UTR 3 NP_001269219.1
NM_174919.3 4403 Intron NP_777579.2
NM_199282.2 4403 UTR 3 NP_954976.1
XM_005257128.3 4403 UTR 3 XP_005257185.1
XM_006721745.2 4403 UTR 3 XP_006721808.1
XM_006721747.2 4403 UTR 3 XP_006721810.1
XM_006721750.3 4403 Intron XP_006721813.1
XM_011524468.1 4403 UTR 3 XP_011522770.1
XM_011524469.1 4403 UTR 3 XP_011522771.1
XM_011524470.2 4403 UTR 3 XP_011522772.1
XM_011524471.1 4403 UTR 3 XP_011522773.1
XM_011524472.2 4403 UTR 3 XP_011522774.1
XM_011524473.2 4403 Intron XP_011522775.1
XM_011524474.1 4403 UTR 3 XP_011522776.1
XM_017024311.1 4403 UTR 3 XP_016879800.1
XM_017024312.1 4403 UTR 3 XP_016879801.1
XM_017024313.1 4403 Intron XP_016879802.1

View Full Product Details