Product Details

SNP ID
rs11012
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:45436075 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CACTACCTTTCTGAGGGAGGGGAGG[C/T]GGGAAGAGTCAATGCATCTGAAAGC
Phenotype
MIM: 610591 MIM: 611466
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP27 PubMed Links

Gene Details

Gene
ARHGAP27
Gene Name
Rho GTPase activating protein 27
There are no transcripts associated with this gene.

Gene
PLEKHM1
Gene Name
pleckstrin homology and RUN domain containing M1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014798.2 4827 UTR 3 NP_055613.1
XM_006722201.3 4827 UTR 3 XP_006722264.1
XM_011525523.2 4827 Intron XP_011523825.1
XM_011525524.1 4827 UTR 3 XP_011523826.1
XM_011525525.1 4827 UTR 3 XP_011523827.1
XM_011525526.1 4827 UTR 3 XP_011523828.1
XM_011525528.2 4827 UTR 3 XP_011523830.1
XM_017025451.1 4827 UTR 3 XP_016880940.1
XM_017025452.1 4827 UTR 3 XP_016880941.1
XM_017025453.1 4827 UTR 3 XP_016880942.1
XM_017025454.1 4827 UTR 3 XP_016880943.1

View Full Product Details