Product Details

SNP ID
rs1777487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:107140968 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAAAATTACTTTGGAGAAAAATC[C/T]GTCCTTCTCACAAACAATATATATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUP62CL PubMed Links

Gene Details

Gene
NUP62CL
Gene Name
nucleoporin 62 C-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017681.2 Intron NP_060151.2

View Full Product Details