Product Details

SNP ID
rs1285582
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:107123872 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAACTAGACAGTAGGTTCCTTTGAG[A/G]AGAAGAGACTTTACAGTATCTAGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUP62CL PubMed Links

Gene Details

Gene
NUP62CL
Gene Name
nucleoporin 62 C-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017681.2 1317 UTR 3 NP_060151.2
Gene
RBM41
Gene Name
RNA binding motif protein 41
There are no transcripts associated with this gene.

View Full Product Details