Product Details
- SNP ID
-
rs1061752
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:47107659 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCTATTTAGTCCCCTTTATTTTGG[G/T]GGGTTTAACATGAGAAGAATAATCC
- Phenotype
-
MIM: 614697
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
HDHD2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114108543] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HDHD2
- Gene Name
- haloacid dehalogenase like hydrolase domain containing 2
- Gene
- KATNAL2
- Gene Name
- katanin catalytic subunit A1 like 2
There are no transcripts associated with this gene.
View Full Product Details