Product Details

SNP ID
rs1136678
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:57972463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCAGCAAGGTCATTCTTGGGCT[C/T]TCCCTACACGAGTCCTTCCAGTAGT
Phenotype
MIM: 606747
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VEZF1 PubMed Links

Gene Details

Gene
VEZF1
Gene Name
vascular endothelial zinc finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007146.2 3923 UTR 3 NP_009077.2
XM_005257643.1 3923 UTR 3 XP_005257700.1
XM_005257644.4 3923 UTR 3 XP_005257701.1
XM_017025017.1 3923 UTR 3 XP_016880506.1
XM_017025018.1 3923 UTR 3 XP_016880507.1

View Full Product Details