Product Details

SNP ID
rs910924
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:752680 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GACAGCAGCGTTCCGGATCCTAGAC[A/G]CTTTTTGACGGCTGGTCCTCCCCAG
Phenotype
MIM: 611627 MIM: 606969
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
DBIL5P PubMed Links
Additional Information
For this assay, SNP(s) [rs57406438] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
DBIL5P
Gene Name
diazepam binding inhibitor-like 5, pseudogene
There are no transcripts associated with this gene.

Gene
FAM57A
Gene Name
family with sequence similarity 57 member A
There are no transcripts associated with this gene.

Gene
GEMIN4
Gene Name
gem nuclear organelle associated protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015721.2 189 Intron NP_056536.2
XM_005256667.4 189 Intron XP_005256724.1
XM_005256670.4 189 UTR 5 XP_005256727.1
XM_011523910.2 189 Intron XP_011522212.1
XM_011523911.2 189 UTR 5 XP_011522213.1
XM_011523912.2 189 Intron XP_011522214.1
XM_011523913.2 189 Intron XP_011522215.1
XM_017024709.1 189 UTR 5 XP_016880198.1
Gene
GLOD4
Gene Name
glyoxalase domain containing 4
There are no transcripts associated with this gene.

View Full Product Details