Product Details

SNP ID
rs1061174
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:46857793 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCCACATGGAAACTAAGCCCCT[C/T]AGCTGAGGGGACAGCTGGAAGAGGA
Phenotype
MIM: 604270
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LRP4 PubMed Links

Gene Details

Gene
LRP4
Gene Name
LDL receptor related protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002334.3 6221 UTR 3 NP_002325.2
XM_011520103.2 6221 UTR 3 XP_011518405.1
XM_011520104.2 6221 UTR 3 XP_011518406.1
XM_017017734.1 6221 Intron XP_016873223.1
Gene
LRP4-AS1
Gene Name
LRP4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details