Product Details

SNP ID
rs1304308
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:149695386 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCACAAAAACACAAATTGATAAA[C/T]TAGATTTCCTCAAACTAAAAACAAA
Phenotype
MIM: 300344
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAGEA11 PubMed Links

Gene Details

Gene
MAGEA11
Gene Name
MAGE family member A11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011544.1 Intron NP_001011544.1
NM_005366.4 Intron NP_005357.2
XM_011531164.2 Intron XP_011529466.1
XM_017029521.1 Intron XP_016885010.1
XM_017029522.1 Intron XP_016885011.1

View Full Product Details