Product Details

SNP ID
rs1476438
Assay Type
Functionally tested
NCBI dbSNP Submissions
47
Location
Chr.1:158932486 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGAAGATAAAATAAGACAAGATAA[C/T]GATGTGCACAAAGTATCAGGAAATA
Phenotype
MIM: 612677
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PYHIN1 PubMed Links

Gene Details

Gene
PYHIN1
Gene Name
pyrin and HIN domain family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152501.4 Intron NP_689714.2
NM_198928.4 Intron NP_945146.1
NM_198929.4 Intron NP_945147.1
NM_198930.3 Intron NP_945148.1
XM_005244930.1 Intron XP_005244987.1
XM_011509242.2 Intron XP_011507544.1
XM_011509243.2 Intron XP_011507545.1
XM_017000463.1 Intron XP_016855952.1

View Full Product Details