Product Details

SNP ID
rs1766923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:94040615 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAATAATAGAATAAATAACATAT[A/G]AAGGAAAAAAGTCAGCCATACTCCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTPDC1 PubMed Links

Gene Details

Gene
PTPDC1
Gene Name
protein tyrosine phosphatase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253829.1 Intron NP_001240758.1
NM_001253830.1 Intron NP_001240759.1
NM_152422.4 Intron NP_689635.3
NM_177995.2 Intron NP_818931.1
XM_005251711.4 Intron XP_005251768.1
XM_011518240.2 Intron XP_011516542.1
XM_011518241.2 Intron XP_011516543.1
XM_017014280.1 Intron XP_016869769.1
XM_017014281.1 Intron XP_016869770.1
XM_017014282.1 Intron XP_016869771.1

View Full Product Details