Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_002441.4 | Intron | NP_002432.1 | |||
NM_025259.5 | Intron | NP_079535.4 | |||
NM_172165.3 | Intron | NP_751897.1 | |||
NM_172166.3 | Intron | NP_751898.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001039651.1 | Intron | NP_001034740.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_025258.2 | Intron | NP_079534.2 | |||
XM_005249427.2 | Intron | XP_005249484.1 | |||
XM_017011327.1 | Intron | XP_016866816.1 | |||
XM_017011328.1 | Intron | XP_016866817.1 | |||
XM_017011329.1 | Intron | XP_016866818.1 |