Product Details

SNP ID
rs351410
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:212285335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCGCGGAAGTTGTCAGTCAAAC[C/T]CCTCCTTCCCACCCCTTGCGAGGCC
Phenotype
MIM: 601643
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101929541 PubMed Links

Gene Details

Gene
LOC101929541
Gene Name
uncharacterized LOC101929541
There are no transcripts associated with this gene.

Gene
PPP2R5A
Gene Name
protein phosphatase 2 regulatory subunit B'alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199756.1 Intron NP_001186685.1
NM_006243.3 Intron NP_006234.1

View Full Product Details