Product Details

SNP ID
rs779805
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:10141653 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCCTAGCCTCGCCTCCGTTACAAC[A/G]GCCTACGGTGCTGGAGGATCCTTCT
Phenotype
MIM: 608537
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
VHL PubMed Links

Gene Details

Gene
VHL
Gene Name
von Hippel-Lindau tumor suppressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000551.3 19 UTR 5 NP_000542.1
NM_198156.2 19 UTR 5 NP_937799.1

View Full Product Details