Product Details

SNP ID
rs13485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17403117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGGGACTCATTGTCCGGAGGGAG[C/G]CTCTGGAGGGAGACAGCCCTGGGTC
Phenotype
MIM: 600534
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BISPR PubMed Links

Gene Details

Gene
BISPR
Gene Name
BST2 interferon stimulated positive regulator (non-protein coding)
There are no transcripts associated with this gene.

Gene
BST2
Gene Name
bone marrow stromal cell antigen 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004335.3 849 UTR 3 NP_004326.1
Gene
LOC105372343
Gene Name
uncharacterized LOC105372343
There are no transcripts associated with this gene.

Gene
MVB12A
Gene Name
multivesicular body subunit 12A
There are no transcripts associated with this gene.

View Full Product Details