Product Details

SNP ID
rs1059369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18386331 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCCCGGGACCCTCAGAGTTGCAC[A/T]CCGAAGACTCCAGATTCCGAGAGTT
Phenotype
MIM: 605312 MIM: 607518
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GDF15 PubMed Links

Gene Details

Gene
GDF15
Gene Name
growth differentiation factor 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004864.2 174 Missense Mutation ACC,TCC T48S NP_004855.2
Gene
LRRC25
Gene Name
leucine rich repeat containing 25
There are no transcripts associated with this gene.

Gene
MIR3189
Gene Name
microRNA 3189
There are no transcripts associated with this gene.

View Full Product Details