Product Details

SNP ID
rs1780705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:37101547 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTACTCTGTGCTCAATCTGTGTAAT[A/G]CCTCTTACATACACTCTTTCATTTA
Phenotype
MIM: 116957
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MROH8 PubMed Links

Gene Details

Gene
MROH8
Gene Name
maestro heat like repeat family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152503.5 Intron NP_689716.4
NM_213631.2 Intron NP_998796.1
NM_213632.2 Intron NP_998797.2
Gene
RBL1
Gene Name
RB transcriptional corepressor like 1
There are no transcripts associated with this gene.

View Full Product Details