Product Details
- SNP ID
-
rs1453561
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:98997461 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCAGATTACAGACAGCTGCCACAGA[C/T]AATAAAATAAAATCGCAATTAAGTT
- Phenotype
-
MIM: 607166
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TSGA10
PubMed Links
Gene Details
- Gene
- TSGA10
- Gene Name
- testis specific 10
View Full Product Details