Product Details

SNP ID
rs1383992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:86608257 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTATTTTAGGAGTAGAGAGATTCGA[C/T]TGTGAAGAATTCACTATGTATAGAA
Phenotype
MIM: 600267
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PTPN13 PubMed Links

Gene Details

Gene
PTPN13
Gene Name
protein tyrosine phosphatase, non-receptor type 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006264.2 Intron NP_006255.1
NM_080683.2 Intron NP_542414.1
NM_080684.2 Intron NP_542415.1
NM_080685.2 Intron NP_542416.1
XM_011532165.1 Intron XP_011530467.1
XM_017008511.1 Intron XP_016864000.1
XM_017008512.1 Intron XP_016864001.1
XM_017008513.1 Intron XP_016864002.1
XM_017008514.1 Intron XP_016864003.1
XM_017008515.1 Intron XP_016864004.1
XM_017008516.1 Intron XP_016864005.1

View Full Product Details