Product Details

SNP ID
rs805259
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31663600 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTAACCCATTGAATCCTAACAAC[A/G]ATTCTTACTACCCCCATTTCTAAGA
Phenotype
MIM: 606907 MIM: 142590 MIM: 115441 MIM: 142610 MIM: 610433
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
APOM PubMed Links
Additional Information
For this assay, SNP(s) [rs149593367] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
APOM
Gene Name
apolipoprotein M
There are no transcripts associated with this gene.

Gene
BAG6
Gene Name
BCL2 associated athanogene 6
There are no transcripts associated with this gene.

Gene
C6orf47
Gene Name
chromosome 6 open reading frame 47
There are no transcripts associated with this gene.

Gene
CSNK2B
Gene Name
casein kinase 2 beta
There are no transcripts associated with this gene.

Gene
GPANK1
Gene Name
G-patch domain and ankyrin repeats 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199237.1 Intron NP_001186166.1
NM_001199238.1 Intron NP_001186167.1
NM_001199239.1 Intron NP_001186168.1
NM_001199240.1 Intron NP_001186169.1
NM_033177.3 Intron NP_149417.1
XM_005249403.3 Intron XP_005249460.1
XM_006715204.1 Intron XP_006715267.1
XM_011514909.1 Intron XP_011513211.1
XM_011514910.1 Intron XP_011513212.1
Gene
LY6G5B
Gene Name
lymphocyte antigen 6 complex, locus G5B
There are no transcripts associated with this gene.

View Full Product Details