Product Details

SNP ID
rs1802073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:37907562 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGACTGGCTGGTTTGGGAGCAGGAG[G/T]CTTTCCCTTTGGTTTGGGGGGATTA
Phenotype
MIM: 607421 MIM: 606570
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NME8 PubMed Links

Gene Details

Gene
NME8
Gene Name
NME/NM23 family member 8
There are no transcripts associated with this gene.

Gene
SFRP4
Gene Name
secreted frizzled related protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003014.3 1344 Missense Mutation ACT,CCT T320P NP_003005.2

View Full Product Details