Product Details

SNP ID
rs1053108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:163453618 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATTGAATATCATACTTATCAAAA[A/G]AAAAAGTTAATTTTCCCCAAAGGCA
Phenotype
MIM: 601578 MIM: 600936
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCNG1 PubMed Links

Gene Details

Gene
CCNG1
Gene Name
cyclin G1
There are no transcripts associated with this gene.

Gene
HMMR
Gene Name
hyaluronan mediated motility receptor
There are no transcripts associated with this gene.

Gene
NUDCD2
Gene Name
NudC domain containing 2
There are no transcripts associated with this gene.

View Full Product Details