Product Details
- SNP ID
-
rs1474920
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:23118346 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAGAGGCTGGCGGTGGGGGCCTGA[G/T]ATCAAGTAGGTCAGCTTGCCACAGG
- Phenotype
-
MIM: 600749
MIM: 604235
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CEBPE
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116791771] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CEBPE
- Gene Name
- CCAAT/enhancer binding protein epsilon
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001805.3 |
|
Intron |
|
|
NP_001796.2 |
- Gene
- SLC7A8
- Gene Name
- solute carrier family 7 member 8
There are no transcripts associated with this gene.
View Full Product Details