Product Details

SNP ID
rs813095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:28677687 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGGGGCAAGGGAGCCAGTGGCCG[A/C]CGACGGGGGACCGGGAAACTTTTCT
Phenotype
MIM: 604444
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
BAMBI PubMed Links

Gene Details

Gene
BAMBI
Gene Name
BMP and activin membrane bound inhibitor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012342.2 193 UTR 5 NP_036474.1

View Full Product Details