Product Details

SNP ID
rs1052581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:153477592 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTCCCCCTCCCCCCACCCCTTCT[C/T]CCACAATGACAAAACTGTAACTCCT
Phenotype
MIM: 612171
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPRM PubMed Links

Gene Details

Gene
RPRM
Gene Name
reprimo, TP53 dependent G2 arrest mediator candidate
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019845.2 1217 UTR 3 NP_062819.1

View Full Product Details