Product Details

SNP ID
rs1334596
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:94413547 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGCTGCTTCAGCCTTTCTTATGCTT[C/T]TTATTTCTGTGGCCAAAATAAGAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TBC1D12 PubMed Links

Gene Details

Gene
TBC1D12
Gene Name
TBC1 domain family member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015188.1 Intron NP_056003.1
XM_006717732.3 Intron XP_006717795.2
XM_006717733.2 Intron XP_006717796.1
XM_011539558.2 Intron XP_011537860.1
XM_017015996.1 Intron XP_016871485.1
XM_017015997.1 Intron XP_016871486.1
XM_017015998.1 Intron XP_016871487.1
XM_017015999.1 Intron XP_016871488.1

View Full Product Details