Product Details

SNP ID
rs1204868
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:17373044 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGTGCCCTGACGCGTGTCTCTTAC[C/T]GGGCAAACCAGGTGTGCCCCCCAGA
Phenotype
MIM: 605347 MIM: 610363
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PADI4 PubMed Links

Gene Details

Gene
PADI4
Gene Name
peptidyl arginine deiminase 4
There are no transcripts associated with this gene.

Gene
PADI6
Gene Name
peptidyl arginine deiminase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207421.4 Intron NP_997304.3

View Full Product Details