Product Details
- SNP ID
-
rs1339065
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:18634032 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCCCAGGTGTGTCCTCATCCCTC[G/T]TAGTGTGGCAGGAGTTGGGGGACAC
- Phenotype
-
MIM: 167410
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PAX7
PubMed Links
Gene Details
- Gene
- PAX7
- Gene Name
- paired box 7
View Full Product Details