Product Details

SNP ID
rs8399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19037817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCCAGCCCCTGCGCTGCTGAGG[A/C]GGGGCCGGGAGGTGTGCATGCGGCC
Phenotype
MIM: 600594
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DGCR11 PubMed Links

Gene Details

Gene
DGCR11
Gene Name
DiGeorge syndrome critical region gene 11 (non-protein coding)
There are no transcripts associated with this gene.

Gene
DGCR2
Gene Name
DiGeorge syndrome critical region gene 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173533.1 2826 UTR 3 NP_001167004.1
NM_001173534.1 2826 UTR 3 NP_001167005.1
NM_001184781.1 2826 UTR 3 NP_001171710.1
NM_005137.2 2826 UTR 3 NP_005128.1
Gene
DGCR5
Gene Name
DiGeorge syndrome critical region gene 5 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details