Product Details
- SNP ID
-
rs1786194
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:118136104 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTCTCAGGTAGGAGGGGGAGAAGC[A/G]GGGGAGAGCTGGGCTCAGGAGTGCC
- Phenotype
-
MIM: 608256
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SCN4B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs59423170] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SCN4B
- Gene Name
- sodium voltage-gated channel beta subunit 4
View Full Product Details