Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001278547.1 | 2014 | UTR 3 | NP_001265476.1 | ||
NM_001278548.1 | 2014 | UTR 3 | NP_001265477.1 | ||
NM_001323302.1 | 2014 | UTR 3 | NP_001310231.1 | ||
NM_001323320.1 | 2014 | UTR 3 | NP_001310249.1 | ||
NM_001323321.1 | 2014 | UTR 3 | NP_001310250.1 | ||
NM_001323322.1 | 2014 | UTR 3 | NP_001310251.1 | ||
NM_001323323.1 | 2014 | UTR 3 | NP_001310252.1 | ||
NM_001323324.1 | 2014 | UTR 3 | NP_001310253.1 | ||
NM_001323325.1 | 2014 | UTR 3 | NP_001310254.1 | ||
NM_001323326.1 | 2014 | UTR 3 | NP_001310255.1 | ||
NM_001323327.1 | 2014 | UTR 3 | NP_001310256.1 | ||
NM_001323328.1 | 2014 | UTR 3 | NP_001310257.1 | ||
NM_001323329.1 | 2014 | UTR 3 | NP_001310258.1 | ||
NM_001323330.1 | 2014 | UTR 3 | NP_001310259.1 | ||
NM_001323331.1 | 2014 | UTR 3 | NP_001310260.1 | ||
NM_139046.3 | 2014 | UTR 3 | NP_620634.1 | ||
NM_139049.3 | 2014 | UTR 3 | NP_620637.1 |