Product Details

SNP ID
rs1628712
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:54641934 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTCATATCTTTGGCTTAGAGCTG[A/G]ATTTTCTGTTACCATGTGGATGCTC
Phenotype
MIM: 606803
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACOT11 PubMed Links

Gene Details

Gene
ACOT11
Gene Name
acyl-CoA thioesterase 11
There are no transcripts associated with this gene.

Gene
MROH7
Gene Name
maestro heat like repeat family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039464.3 195 UTR 5 NP_001034553.2
NM_001291332.1 195 UTR 5 NP_001278261.1
Gene
MROH7-TTC4
Gene Name
MROH7-TTC4 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details