Product Details

SNP ID
rs1779047
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:37772715 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGAATCCCTTTAGTCACGTATTT[G/T]CTCCCTGTTGATAACCTGACAGGGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF248 PubMed Links

Gene Details

Gene
ZNF248
Gene Name
zinc finger protein 248
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267597.1 Intron NP_001254526.1
NM_001267605.1 Intron NP_001254534.1
NM_001267606.1 Intron NP_001254535.1
NM_001267607.1 Intron NP_001254536.1
NM_021045.2 Intron NP_066383.1
XM_006717485.3 Intron XP_006717548.1
XM_011519597.2 Intron XP_011517899.1
XM_011519598.2 Intron XP_011517900.1
XM_011519599.1 Intron XP_011517901.1
XM_011519600.2 Intron XP_011517902.1
XM_017016444.1 Intron XP_016871933.1
XM_017016445.1 Intron XP_016871934.1
XM_017016446.1 Intron XP_016871935.1
XM_017016447.1 Intron XP_016871936.1
XM_017016448.1 Intron XP_016871937.1
XM_017016449.1 Intron XP_016871938.1
XM_017016450.1 Intron XP_016871939.1
XM_017016451.1 Intron XP_016871940.1

View Full Product Details