Product Details

SNP ID
rs1322316
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:37762832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTTTCTTACTAAGCATTTTATGG[A/G]ATAGTACAATGAGCATTACCATTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF248 PubMed Links

Gene Details

Gene
ZNF248
Gene Name
zinc finger protein 248
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267597.1 Intron NP_001254526.1
NM_001267605.1 Intron NP_001254534.1
NM_001267606.1 Intron NP_001254535.1
NM_021045.2 Intron NP_066383.1

View Full Product Details