Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001267597.1 | Intron | NP_001254526.1 | |||
NM_001267605.1 | Intron | NP_001254534.1 | |||
NM_001267606.1 | Intron | NP_001254535.1 | |||
NM_001267607.1 | Intron | NP_001254536.1 | |||
NM_021045.2 | Intron | NP_066383.1 | |||
XM_006717485.3 | Intron | XP_006717548.1 | |||
XM_011519597.2 | Intron | XP_011517899.1 | |||
XM_011519598.2 | Intron | XP_011517900.1 | |||
XM_011519599.1 | Intron | XP_011517901.1 | |||
XM_011519600.2 | Intron | XP_011517902.1 | |||
XM_017016444.1 | Intron | XP_016871933.1 | |||
XM_017016445.1 | Intron | XP_016871934.1 | |||
XM_017016446.1 | Intron | XP_016871935.1 | |||
XM_017016447.1 | Intron | XP_016871936.1 | |||
XM_017016448.1 | Intron | XP_016871937.1 | |||
XM_017016449.1 | Intron | XP_016871938.1 | |||
XM_017016450.1 | Intron | XP_016871939.1 | |||
XM_017016451.1 | Intron | XP_016871940.1 |