Product Details

SNP ID
rs876541
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:20140790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACACTGGAGAAAGCAAACAGGCTG[C/T]CCACAAAGAATGATTAGGAAGCTTG
Phenotype
MIM: 121015
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GJA3 PubMed Links

Gene Details

Gene
GJA3
Gene Name
gap junction protein alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021954.3 2578 UTR 3 NP_068773.2
XM_011535048.2 2578 UTR 3 XP_011533350.1

View Full Product Details