Product Details

SNP ID
rs1561303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:53849257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATATACATGTGTACATACACAGG[C/T]ACCTGGAGTGGTCCCACGTGCTCTG
Phenotype
MIM: 605458
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHDH PubMed Links

Gene Details

Gene
CHDH
Gene Name
choline dehydrogenase
There are no transcripts associated with this gene.

Gene
IL17RB
Gene Name
interleukin 17 receptor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018725.3 Intron NP_061195.2
XM_005265310.4 Intron XP_005265367.1
XM_005265311.4 Intron XP_005265368.1
XM_005265312.4 Intron XP_005265369.1
XM_011533941.2 Intron XP_011532243.1
XM_017006804.1 Intron XP_016862293.1
XM_017006805.1 Intron XP_016862294.1
XM_017006806.1 Intron XP_016862295.1
XM_017006807.1 Intron XP_016862296.1

View Full Product Details