Product Details
- SNP ID
-
rs796860
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
68
- Location
-
Chr.1:183555720 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGGGTGTCTTGTTTTTGTAAGATG[A/C]TAGGTTTTTTTTTATTGTGGTATGA
- Phenotype
-
MIM: 608515
MIM: 610964
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NCF2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs543935249] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NCF2
- Gene Name
- neutrophil cytosolic factor 2
- Gene
- SMG7
- Gene Name
- SMG7, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.
View Full Product Details