Product Details

SNP ID
rs1557727
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:14003416 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCAGTCTCCTCATCTGCAAAATGA[C/G]AATAATATTCCTTCAACACATTTCT
Phenotype
MIM: 300962
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GEMIN8 PubMed Links

Gene Details

Gene
GEMIN8
Gene Name
gem nuclear organelle associated protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042479.1 Intron NP_001035944.1
NM_001042480.1 Intron NP_001035945.1
NM_017856.2 Intron NP_060326.1
XM_005274555.2 Intron XP_005274612.1
XM_017029617.1 Intron XP_016885106.1
XM_017029618.1 Intron XP_016885107.1

View Full Product Details