Product Details

SNP ID
rs1107655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:185080780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGAGCCATGCCCTGGCAGGGGG[A/C]AGCTCCTCATGCTCACACCTGCCTC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C3orf70 PubMed Links

Gene Details

Gene
C3orf70
Gene Name
chromosome 3 open reading frame 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025266.1 3171 UTR 3 NP_001020437.1

View Full Product Details