Product Details

SNP ID
rs1468502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:12022796 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAAGTAATAAAGGTAACAAGCTG[C/G]GTTTTGTGATAGAAGATAATAGAGG
Phenotype
MIM: 601335
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAP2K4 PubMed Links

Gene Details

Gene
MAP2K4
Gene Name
mitogen-activated protein kinase kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281435.1 Intron NP_001268364.1
NM_003010.3 Intron NP_003001.1
XM_005256753.2 Intron XP_005256810.1
XM_005256755.2 Intron XP_005256812.1
XM_011523976.2 Intron XP_011522278.1
XM_011523977.2 Intron XP_011522279.1

View Full Product Details